Assay Design
Gene Transcripts
- Gene Symbol
- Csn1s2b
- Entrez Gene ID
-
286759
- Gene Name
- casein alpha s2-like B
- Gene Aliases
-
Csnd
- Chromosome Location
-
Chr.14: 21899954 - 21916068 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173106.1 |
NP_775129.1 |
- |
229 |
67 |
GenBank mRNA |
AY154894.1 |
- |
- |
229 |
67 |
View Full Product Details