Assay Design



Gene Transcripts

Gene Symbol
Vma21
Entrez Gene ID
501658
Gene Name
VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Gene Aliases
RGD1566155
Chromosome Location
Chr.16: 70142561 - 70151838 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253301.2 XP_006253363.1 - 299 112
XM_008771344.1 XP_008769566.1 - 570 112
GenBank mRNA FQ211836.1 - - 263 112
FQ215711.1 - - 565 112

Gene Symbol
LOC100912478
Entrez Gene ID
100912478
Gene Name
vacuolar ATPase assembly integral membrane protein VMA21-like
Gene Aliases
-
Chromosome Location
Chr.17: 35127078 - 35134500 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751684.3 XP_003751732.1 2 - 3 299 112
XM_006253887.2 XP_006253949.1 3 - 4 570 112

Gene Symbol
LOC103689964
Entrez Gene ID
103689964
Gene Name
vacuolar ATPase assembly integral membrane protein Vma21
Gene Aliases
-
Chromosome Location
Chr.17: 34940424 - 34946014 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253890.2 XP_006253952.1 - 570 112
XM_577054.6 XP_577054.2 - 299 112

Gene Symbol
LOC103694079
Entrez Gene ID
103694079
Gene Name
vacuolar ATPase assembly integral membrane protein Vma21-like
Gene Aliases
-
Chromosome Location
Chr.17: 35121571 - 35126408 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771606.1 XP_008769828.1 - 133 112

View Full Product Details