Assay Design



Gene Transcripts

Gene Symbol
Eepd1
Entrez Gene ID
315500
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
RGD1307401
Chromosome Location
Chr.8: 26651264 - 26758330 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242726.2 XP_006242788.1 1 - 2 323 146
XM_006242727.2 XP_006242789.1 1 - 2 323 146
GenBank mRNA BC099214.1 - 1 - 2 281 146

View Full Product Details