Assay Design



Gene Transcripts

Gene Symbol
Eepd1
Entrez Gene ID
315500
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
RGD1307401
Chromosome Location
Chr.8: 26651264 - 26758330 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014088.3 NP_001014110.1 4 - 5 1226 62
XM_006242726.2 XP_006242788.1 5 - 6 1660 62
XM_006242727.2 XP_006242789.1 4 - 5 1549 62
GenBank mRNA BC083816.1 - 4 - 5 1226 62
BC099214.1 - 5 - 6 1618 62

View Full Product Details