Assay Design



Gene Transcripts

Gene Symbol
Kcnd1
Entrez Gene ID
116695
Gene Name
potassium channel, voltage gated Shal-related subfamily D, member 1
Gene Aliases
Kv4.1
Chromosome Location
Chr.X: 15506724 - 15520712 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105748.1 NP_001099218.1 3 - 4 1369 80

View Full Product Details