Assay Design



Gene Transcripts

Gene Symbol
Brix1
Entrez Gene ID
294799
Gene Name
BRX1, biogenesis of ribosomes
Gene Aliases
Bxdc2, RGD1308508
Chromosome Location
Chr.2: 60450547 - 60461425 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029915.1 NP_001025086.1 7 - 8 565 83
GenBank mRNA BC098005.1 - 7 - 8 565 83

View Full Product Details