Assay Design



Gene Transcripts

Gene Symbol
C1qtnf3
Entrez Gene ID
294806
Gene Name
C1q and tumor necrosis factor related protein 3
Gene Aliases
-
Chromosome Location
Chr.2: 60920101 - 60942586 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134436.1 NP_001127908.1 2 - 3 479 95
XM_006232067.2 XP_006232129.2 2 - 3 416 95
GenBank mRNA BC162048.1 - 2 - 3 479 95

View Full Product Details