Assay Design



Gene Transcripts

Gene Symbol
Cdc40
Entrez Gene ID
361859
Gene Name
cell division cycle 40
Gene Aliases
-
Chromosome Location
Chr.20: 48503973 - 48574546 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108538.1 NP_001102008.1 7 - 8 1066 65
XM_008773000.1 XP_008771222.1 9 - 10 1036 65
GenBank mRNA BC166724.1 - 9 - 10 997 65

View Full Product Details