Assay Design



Gene Transcripts

Gene Symbol
Ddah2
Entrez Gene ID
294239
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
-
Chromosome Location
Chr.20: 5049265 - 5052573 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165936.1 NP_001159408.1 1 - 2 136 101
GenBank mRNA EV770973.1 - 1 - 2 136 101

View Full Product Details