Assay Design



Gene Transcripts

Gene Symbol
Gtf2h5
Entrez Gene ID
502227
Gene Name
general transcription factor IIH, polypeptide 5
Gene Aliases
RGD1560991
Chromosome Location
Chr.1: 46978340 - 46985032 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126088.1 NP_001119560.1 2 - 3 222 110
XM_006227894.1 XP_006227956.1 2 - 3 183 110
GenBank mRNA FQ212207.1 - - 144 110

View Full Product Details