Assay Design



Gene Transcripts

Gene Symbol
Ezr
Entrez Gene ID
54319
Gene Name
ezrin
Gene Aliases
Vil2
Chromosome Location
Chr.1: 47287872 - 47331412 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019357.1 NP_062230.1 1 - 2 497 65
GenBank mRNA AF450298.1 - 1 - 2 96 65
AY428869.1 - 1 - 2 82 65
BC081958.1 - 1 - 2 497 65

View Full Product Details