Assay Design



Gene Transcripts

Gene Symbol
Samd9l
Entrez Gene ID
500015
Gene Name
sterile alpha motif domain containing 9-like
Gene Aliases
-
Chromosome Location
Chr.4: 31296710 - 31299424 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236084.2 XP_006236146.2 1 - 1 1444 133
GenBank mRNA BC098747.1 - - 1436 133

View Full Product Details