Assay Design



Gene Transcripts

Gene Symbol
Hdhd2
Entrez Gene ID
361351
Gene Name
haloacid dehalogenase-like hydrolase domain containing 2
Gene Aliases
Ier3ip1, LRRG00122, RGD1308579
Chromosome Location
Chr.18: 73226630 - 73278062 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014151.2 NP_001014173.2 1 - 2 95 98
GenBank mRNA AY539873.1 - 1 - 2 95 98

View Full Product Details