Assay Design



Gene Transcripts

Gene Symbol
Il23a
Entrez Gene ID
155140
Gene Name
Interleukin 23, alpha subunit p19
Gene Aliases
-
Chromosome Location
Chr.7: 2710609 - 2712723 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130410.2 NP_569094.1 2 - 3 572 58
GenBank mRNA AY055379.1 - 2 - 3 300 58
BC098907.1 - 2 - 3 572 58

View Full Product Details