Assay Design



Gene Transcripts

Gene Symbol
Hiatl1
Entrez Gene ID
306687
Gene Name
hippocampus abundant transcript-like 1
Gene Aliases
RGD1308377
Chromosome Location
Chr.17: 2607777 - 2660902 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107334.1 NP_001100804.1 1 - 2 469 94
GenBank mRNA BC166784.1 - 1 - 2 462 94
FQ230882.1 - 1 - 2 474 94

View Full Product Details