Assay Design



Gene Transcripts

Gene Symbol
Prss47
Entrez Gene ID
680378
Gene Name
protease, serine, 47
Gene Aliases
-
Chromosome Location
Chr.17: 2814264 - 2828355 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253500.2 XP_006253562.1 2 - 3 785 120
XM_008771453.1 XP_008769675.1 2 - 3 786 120
XM_008771454.1 XP_008769676.1 2 - 3 508 120

View Full Product Details