Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001073777.4 | XP_001073777.2 | 14 - 15 | 1844 | 80 |
XM_006244268.2 | XP_006244330.1 | 15 - 16 | 1910 | 80 | |
XM_006244269.2 | XP_006244331.1 | 15 - 16 | 2139 | 80 | |
XM_006244270.2 | XP_006244332.1 | 15 - 16 | 2424 | 80 | |
XM_006244271.2 | XP_006244333.1 | 15 - 16 | 2436 | 80 | |
XM_008766793.1 | XP_008765015.1 | 14 - 15 | 2269 | 80 | |
XM_008766794.1 | XP_008765016.1 | 11 - 12 | 2528 | 80 |