Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013957.1 | NP_001013979.1 | 3 - 4 | 327 | 67 |
XM_006256707.1 | XP_006256769.1 | 3 - 4 | 301 | 67 | |
XM_006256708.2 | XP_006256770.1 | 2 - 3 | 202 | 67 | |
GenBank mRNA | BC059163.1 | - | 3 - 4 | 327 | 67 |