Assay Design



Gene Transcripts

Gene Symbol
Was
Entrez Gene ID
317371
Gene Name
Wiskott-Aldrich syndrome
Gene Aliases
WASP
Chromosome Location
Chr.X: 15155246 - 15164099 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108248.1 NP_001101718.1 6 - 7 585 141
GenBank mRNA BC158677.1 - 6 - 7 601 141

View Full Product Details