Assay Design



Gene Transcripts

Gene Symbol
Lhfpl1
Entrez Gene ID
300286
Gene Name
lipoma HMGIC fusion partner-like 1
Gene Aliases
Lhfp
Chromosome Location
Chr.X: 116577283 - 116638637 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181085.2 NP_851599.1 1 - 2 151 85
XM_008773405.1 XP_008771627.1 2 - 3 184 85
GenBank mRNA AY253667.1 - 1 - 2 151 85

View Full Product Details