Assay Design



Gene Transcripts

Gene Symbol
Rbm38
Entrez Gene ID
366262
Gene Name
RNA binding motif protein 38
Gene Aliases
Rnpc1
Chromosome Location
Chr.3: 171037896 - 171050500 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108965.1 NP_001102435.1 1 - 2 1022 97
XM_006235704.2 XP_006235766.1 2 - 3 492 97
XM_008762528.1 XP_008760750.1 3 - 4 582 97

View Full Product Details