Assay Design



Gene Transcripts

Gene Symbol
Dok5
Entrez Gene ID
502694
Gene Name
docking protein 5
Gene Aliases
RGD1562846
Chromosome Location
Chr.3: 168345117 - 168482445 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109344.2 NP_001102814.1 1 - 2 139 110
XM_006235673.2 XP_006235735.1 1 - 2 318 110
GenBank mRNA BC167027.1 - 1 - 2 139 110

View Full Product Details