Assay Design



Gene Transcripts

Gene Symbol
Cstf1
Entrez Gene ID
311670
Gene Name
cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa
Gene Aliases
-
Chromosome Location
Chr.3: 170380414 - 170392162 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235683.2 XP_006235745.1 1 - 2 166 63

View Full Product Details