Assay Design



Gene Transcripts

Gene Symbol
Pmel
Entrez Gene ID
362818
Gene Name
premelanosome protein
Gene Aliases
Si, Silv
Chromosome Location
Chr.7: 3133343 - 3143827 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240779.2 XP_006240841.1 1 - 2 585 62

View Full Product Details