Assay Design



Gene Transcripts

Gene Symbol
Scgn
Entrez Gene ID
306942
Gene Name
secretagogin, EF-hand calcium binding protein
Gene Aliases
-
Chromosome Location
Chr.17: 43357374 - 43399726 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771653.1 XP_008769875.1 7 - 8 876 148

View Full Product Details