Assay Design



Gene Transcripts

Gene Symbol
Abt1
Entrez Gene ID
306960
Gene Name
activator of basal transcription 1
Gene Aliases
-
Chromosome Location
Chr.17: 43946122 - 43948224 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025674.1 NP_001020845.1 1 - 2 291 64
GenBank mRNA BC099109.1 - 1 - 2 291 64

View Full Product Details