Assay Design



Gene Transcripts

Gene Symbol
Tdp2
Entrez Gene ID
498749
Gene Name
tyrosyl-DNA phosphodiesterase 2
Gene Aliases
Ttrap
Chromosome Location
Chr.17: 42229667 - 42241025 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034947.1 NP_001030119.1 2 - 3 337 66
GenBank mRNA BC101920.1 - 2 - 3 337 66

View Full Product Details