Assay Design



Gene Transcripts

Gene Symbol
Slc17a3
Entrez Gene ID
266730
Gene Name
solute carrier family 17 (organic anion transporter), member 3
Gene Aliases
Rnpt4
Chromosome Location
Chr.17: 43518760 - 43545159 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153622.2 NP_705886.1 12 - 13 1580 122
XM_006253941.2 XP_006254003.1 15 - 16 2233 122
XM_006253944.2 XP_006254006.1 15 - 16 2222 122
XM_006253946.2 XP_006254008.1 14 - 15 2109 122
XM_006253948.2 XP_006254010.1 10 - 11 2110 122
XM_006253949.2 XP_006254011.1 13 - 14 1992 122
XM_006253950.2 XP_006254012.1 14 - 15 2002 122
XM_006253951.2 XP_006254013.1 13 - 14 1876 122
XM_008771623.1 XP_008769845.1 16 - 17 2310 122
XM_008771624.1 XP_008769846.1 16 - 17 2298 122
XM_008771625.1 XP_008769847.1 14 - 15 2068 122
GenBank mRNA AB025223.1 - 12 - 13 1604 122
AB025224.1 - 11 - 12 1517 122

View Full Product Details