Assay Design



Gene Transcripts

Gene Symbol
Tex9
Entrez Gene ID
300822
Gene Name
testis expressed 9
Gene Aliases
-
Chromosome Location
Chr.8: 79083449 - 79120391 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729938.4 XP_002729984.1 7 - 8 1560 90
XM_006243344.2 XP_006243406.1 7 - 8 1527 90

View Full Product Details