Assay Design



Gene Transcripts

Gene Symbol
Pigb
Entrez Gene ID
315807
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class B
Gene Aliases
-
Chromosome Location
Chr.8: 79691407 - 79715284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108166.3 NP_001101636.3 10 - 11 1412 85

View Full Product Details