Assay Design



Gene Transcripts

Gene Symbol
Rab8b
Entrez Gene ID
266688
Gene Name
RAB8B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.8: 72641680 - 72714646 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153317.1 NP_695229.1 7 - 8 597 117
GenBank mRNA BC158579.1 - 7 - 8 623 117
U53475.1 - 7 - 8 597 117

View Full Product Details