Assay Design



Gene Transcripts

Gene Symbol
Tmc4
Entrez Gene ID
308310
Gene Name
transmembrane channel-like 4
Gene Aliases
-
Chromosome Location
Chr.1: 64114687 - 64126576 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034104.1 NP_001029276.1 - 2057 110
GenBank mRNA BC100657.1 - - 2057 110

View Full Product Details