Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183332.1 | NP_899161.1 | 2 - 3 | 184 | 63 |
XM_006228035.2 | XP_006228097.1 | 2 - 3 | 375 | 63 | |
GenBank mRNA | AY344060.1 | - | 2 - 3 | 184 | 63 |
BC087054.1 | - | 1 - 2 | 95 | 63 | |
BC107558.1 | - | 1 - 2 | 196 | 63 | |
BC128719.1 | - | 2 - 3 | 341 | 63 |