Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105994.1 | NP_001099464.1 | 6 - 7 | 812 | 181 |
XM_006250541.1 | XP_006250603.1 | 6 - 7 | 865 | 181 | |
XM_008769927.1 | XP_008768149.1 | 6 - 7 | 1724 | 181 | |
GenBank mRNA | FQ212126.1 | - | 6 - 7 | 836 | 181 |
FQ213206.1 | - | 6 - 7 | 738 | 181 |