Assay Design



Gene Transcripts

Gene Symbol
Tmed11
Entrez Gene ID
689712
Gene Name
transmembrane emp24 protein transport domain containing 11
Gene Aliases
-
Chromosome Location
Chr.14: 1936972 - 1957860 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001071744.5 XP_001071744.1 3 - 4 417 113

View Full Product Details