Assay Design
Gene Transcripts
- Gene Symbol
- Ahcy
- Entrez Gene ID
-
29443
- Gene Name
- adenosylhomocysteinase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 150587833 - 150603057 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_017201.1 |
NP_058897.1 |
8 - 9 |
993 |
62 |
GenBank mRNA |
M15185.1 |
- |
8 - 9 |
993 |
62 |
View Full Product Details