Assay Design



Gene Transcripts

Gene Symbol
Rspry1
Entrez Gene ID
689249
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
RGD1308847
Chromosome Location
Chr.19: 10770572 - 10826895 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100945.1 NP_001094415.1 11 - 12 1736 101
GenBank mRNA AY724539.1 - 10 - 11 1379 101
BC129112.1 - 11 - 12 1736 101

View Full Product Details