Assay Design



Gene Transcripts

Gene Symbol
Large
Entrez Gene ID
361368
Gene Name
like-glycosyltransferase
Gene Aliases
-
Chromosome Location
Chr.19: 12481563 - 12943594 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108439.1 NP_001101909.1 12 - 13 2217 58
XM_006255140.2 XP_006255202.1 14 - 15 3188 58

View Full Product Details