Assay Design



Gene Transcripts

Gene Symbol
Npc1
Entrez Gene ID
266732
Gene Name
Niemann-Pick disease, type C1
Gene Aliases
Cdig2
Chromosome Location
Chr.18: 3616878 - 3662715 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153624.2 NP_705888.2 15 - 16 2467 83
XM_006254403.2 XP_006254465.1 15 - 16 2523 83

View Full Product Details