Assay Design



Gene Transcripts

Gene Symbol
Mrps18b
Entrez Gene ID
294230
Gene Name
mitochondrial ribosomal protein S18B
Gene Aliases
Ptd017
Chromosome Location
Chr.20: 3344870 - 3350929 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212534.2 NP_997699.2 5 - 6 439 72
GenBank mRNA BC166875.1 - 5 - 6 421 72
FQ215273.1 - 5 - 6 412 72

View Full Product Details