Assay Design



Gene Transcripts

Gene Symbol
Atat1
Entrez Gene ID
361789
Gene Name
alpha tubulin acetyltransferase 1
Gene Aliases
3110080J08Rik, Mec17, RGD1303066, TAT
Chromosome Location
Chr.20: 3351268 - 3364565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212498.1 NP_997663.1 2 - 3 187 62
XM_006255948.2 XP_006256010.1 2 - 3 222 62
XM_006255950.2 XP_006256012.1 2 - 3 222 62
XM_006255951.2 XP_006256013.1 2 - 3 222 62
XM_008772731.1 XP_008770953.1 2 - 3 222 62

View Full Product Details