Assay Design



Gene Transcripts

Gene Symbol
Atat1
Entrez Gene ID
361789
Gene Name
alpha tubulin acetyltransferase 1
Gene Aliases
3110080J08Rik, Mec17, RGD1303066, TAT
Chromosome Location
Chr.20: 3351268 - 3364565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212498.1 NP_997663.1 8 - 9 670 72
XM_006255950.2 XP_006256012.1 8 - 9 705 72

View Full Product Details