Assay Design



Gene Transcripts

Gene Symbol
Krt80
Entrez Gene ID
315318
Gene Name
keratin 80, type II
Gene Aliases
Kb2, Kb20
Chromosome Location
Chr.7: 142995715 - 143015932 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008815.1 NP_001008815.1 4 - 5 665 73

View Full Product Details