Assay Design



Gene Transcripts

Gene Symbol
Amhr2
Entrez Gene ID
29530
Gene Name
anti-Mullerian hormone receptor, type II
Gene Aliases
-
Chromosome Location
Chr.7: 144052202 - 144060678 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030998.1 NP_112260.1 3 - 4 503 102
GenBank mRNA KJ923229.1 - 3 - 4 503 102
U42427.1 - 3 - 4 503 102
X71916.1 - 3 - 4 485 102

View Full Product Details