Assay Design



Gene Transcripts

Gene Symbol
Aaas
Entrez Gene ID
300259
Gene Name
achalasia, adrenocortical insufficiency, alacrimia
Gene Aliases
-
Chromosome Location
Chr.7: 143937198 - 143956668 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106795.1 NP_001100265.1 15 - 16 1573 127
XM_006242387.2 XP_006242449.1 14 - 15 1490 127
XM_008765704.1 XP_008763926.1 14 - 15 1533 127
XM_008765705.1 XP_008763927.1 14 - 15 1514 127

View Full Product Details