Assay Design



Gene Transcripts

Gene Symbol
Myg1
Entrez Gene ID
300258
Gene Name
melanocyte proliferating gene 1
Gene Aliases
C12orf10
Chromosome Location
Chr.7: 143929662 - 143936865 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005545.1 NP_001005545.1 6 - 7 954 71
GenBank mRNA BC082112.1 - 6 - 7 954 71
FQ230067.1 - 6 - 7 984 71

View Full Product Details