Assay Design



Gene Transcripts

Gene Symbol
Lima1
Entrez Gene ID
300228
Gene Name
LIM domain and actin binding 1
Gene Aliases
eplin
Chromosome Location
Chr.7: 141428113 - 141528445 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191615.2 NP_001178544.1 6 - 7 963 59
XM_006257354.2 XP_006257416.1 7 - 8 1428 59
XM_006257355.2 XP_006257417.1 7 - 8 1111 59

View Full Product Details