Assay Design



Gene Transcripts

Gene Symbol
Smarcd1
Entrez Gene ID
363002
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
Gene Aliases
-
Chromosome Location
Chr.7: 141356185 - 141366725 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108752.1 NP_001102222.1 2 - 3 369 100

View Full Product Details