Assay Design



Gene Transcripts

Gene Symbol
Dennd2c
Entrez Gene ID
295333
Gene Name
DENN/MADD domain containing 2C
Gene Aliases
RGD1308197
Chromosome Location
Chr.2: 205592352 - 205660619 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191569.1 NP_001178498.1 4 - 5 1319 82
XM_006233069.1 XP_006233131.1 4 - 5 1310 82
XM_006233070.2 XP_006233132.1 4 - 5 1637 82
XM_006233071.2 XP_006233133.1 4 - 5 1422 82

View Full Product Details