Assay Design



Gene Transcripts

Gene Symbol
Capza1
Entrez Gene ID
691149
Gene Name
capping protein (actin filament) muscle Z-line, alpha 1
Gene Aliases
-
Chromosome Location
Chr.2: 207305748 - 207350812 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109625.2 NP_001103095.2 9 - 10 1009 107
GenBank mRNA BC166464.1 - 9 - 10 724 107
CB557114.1 - 5 - 6 478 107

View Full Product Details